Procedure
  1. Extract the contents of the UpdaterPackage.zip file to an empty folder, for example, C:\UpdaterPackage.
  2. Modify the InstallResponseFile.xml file to customize the installation.
  3. IMPORTANT
    Do not modify any other file.
  4. Open the Chromeleon Administration Console.
  5. Select Domain Resources > Computers.
  6. Select Create Updater Package (or, for later versions, Installation and Maintenance > Updater > Create Updater Package).
  7. Select the new folder from step 1 as the Package Source.
  8. Select Next.
  9. Select a package name.
  10. Select Next.
  11. Select Finish.