To verify that WebAPI Service has been installed, do the following:
Procedure
- Copy the following link into a browser of your choice: https://localhost:8082/services/ChromeleonServer/Status.
- If you are not using secure transfer, change https to http.
- The link includes the default web API port. If you changed the web API port for your installation, replace the default number with the new number.
- If you are using an SSL certificate, replace the server name localhost with the name of the SSL certificate.
- The installed version is displayed. Example: