You can install the WebAPI Service on top of an existing SampleManager Chromeleon Link Legacy installation. The Windows Service of the Legacy web API is automatically disabled during installation of the WebAPI Service. There are options to use the WebAPI Service and the Legacy web API in parallel. You can always revert to the original Legacy installation by uninstalling the WebAPI Service.