Known issues and limitations are listed in the following table. The numbers in the first column of the table refer to the Thermo Fisher Scientific tracking IDs.
ID
Description
760168
The system supports the use of multiple Chromeleon user accounts to modify sequences and control instruments, ensuring proper documentation of user actions and privileges. However, sending commands to the service with different user credentials in rapid succession may decrease performance.