Ensure the following conditions are met before installing the WebAPI Service:
- SampleManager LIMS version 21.0 or later is installed and running correctly.
- Chromeleon CDS version 7.2.10 ES MUg or later is installed and running correctly.
- Refer to the SampleManager LIMS Release Notes for the required version of the Chromeleon CDS for the respective WebAPI Service functionality.