Nonlinear response and loss of sensitivity may occur from carbonate contamination. Thermo Fisher Scientific recommends a freshly prepared 1M base treatment under these conditions for the Eluent Suppressor component. Wear appropriate safety gear during this operation.
Procedure
- Disconnect the lines to the Eluent Suppressor and perform the following treatment on the Eluent Suppressor component.
- Push 3 mL of 1 M NaOH using a syringe through ELUENT IN port and divert a line out of ELUENT OUT port to waste.
- Push 5 mL of 1 M NaOH using a syringe through REGEN IN and divert a line out of REGEN OUT port to waste.
- Allow the Eluent Suppressor unit to equilibrate for 20 minutes
- Repeat steps 2 and 3 with DI water and reinstall the unit on the system.
- Repeat treatment for a longer time if problem persists.