When using these detectors, the power setting will be shown on the screen when the ERS is on. The SRS/ERS cable must be connected to the ERS and conductive liquid must be flowing to prevent an alarm state.
When the SRS/ERS Control unit is in shutdown mode after detecting a malfunction in the system, an alarm message “SRS Alarm” will flash on the screen. To determine the cause, follow the steps below.
Procedure
- Press Menu, then navigate to Diagnostics Menu > Analog Status.
- On the left side of the screen, two fields give the SRS/ERS status:
- SRS CONNECTED: Y (or N): This indicates that the Dionex SC-CERS 500 is connected. If it shows “N,” connect the SRS/ERS cable.
- SRS OVER VOLT: N (or Y): This indicates whether the Dionex SC-CERS 500 has a voltage higher than the upper limit (7.5 to 8 V). Lower the power setting on the front panel.