This section provides solutions for operating problems that might arise while using the Dionex SC-CERS 500. For more information on problems that originate with the Ion Chromatograph or the specific cation exchange column set in use, refer to the Troubleshooting Guide in the appropriate Installation Manual.
If you cannot solve the problem on your own, contact the Thermo Scientific Regional Office nearest you.
CAUTION
Do not allow eluent to flow through the Dionex SC-CERS 500 without the power turned on. Doing so will cause noticeable anomalous analyte peak areas. If this should occur, perform the procedure outlined in Nonlinear response or loss of sensitivity.