The Dionex SC-CERS 500 device will give a higher background than a standard CERS 500 suppressor for a given application. This is due to the fact that the device transfers MSA across the membranes in order to aid dissociation of weak bases. This transfer increases the background.
For a system operating under optimal conditions:
- Using CS12A operated with 20 mM MSA, the expected background should be less than 5 μS/cm and the peak to peak noise < 3 nS/cm.
- Using CS16 operated with 30 mM MSA at 65°C, the expected background should be less than 7 μS/cm and peak to peak noise < 4 nS/cm.
The following table shows the expected backgrounds at various concentrations.
Eluent Concentration | Expected Background (4 mm) | Expected Background (2 mm) |
|---|---|---|
≤20 mM | < 5 µS/cm | < 5 µS/cm |
20 – 30 mM | < 7 µS/cm | < 7 µS/cm |
30 – 40 mM | < 15 µS/cm | < 10 µS/cm |
40 – 50 mM | < 40 µS/cm | < 10 µS/cm |
Some differences in background may occur dependent on the ambient temperature of the laboratory. Also note that the noise will increase with increasing background.