NOTE
Dionex SC-CERS 500 applications will operate best at current settings of 300 mA or lower. Operating the Dionex SC-CERS 500 4- mm at current settings over 300 mA or Dionex SC-CERS 500 2 mm at current settings over 100 mA can reduce the life of the suppressor.
Eluent Concentration, Methanesulfonic Acid, mM | Eluent Flowrate | Optimized current setting (mA) | Current Selection with older systems (mA) |
|---|---|---|---|
9 | 0.25 | 7 | 50 |
9 | 1.00 | 27 | 50 |
10 | 0.25 | 8 | 50 |
10 | 0.50 | 15 | 50 |
10 | 1.00 | 30 | 50 |
10 | 1.25 | 37 | 50 |
10 | 2.00 | 59 | 100 |
16 | 0.50 | 24 | 50 |
16 | 2.00 | 95 | 100 |
18 | 1.00 | 53 | 100 |
18 | 0.50 | 27 | 50 |
20 | 0.25 | 15 | 50 |
20 | 0.50 | 30 | 50 |
20 | 1.00 | 59 | 100 |
22 | 0.25 | 17 | 50 |
22 | 0.50 | 33 | 50 |
25 | 0.36 | 27 | 50 |
26 | 1.00 | 77 | 100 |
28 | 1.00 | 82 | 100 |
30 | 0.36 | 32 | 50 |
30 | 1.00 | 88 | 100 |
33 | 0.50 | 49 | 50 |
33 | 0.80 | 78 | 100 |
33 | 1.00 | 97 | 100 |
34 | 1.00 | 100 | 100 |
35 | 0.25 | 26 | 50 |
35 | 1.00 | 103 | 300 |
36 | 1.00 | 106 | 300 |
38 | 0.36 | 41 | 50 |
40 | 0.25 | 30 | 50 |
40 | 1.00 | 117 | 300 |
44 | 1.00 | 129 | 300 |
48 | 1.00 | 141 | 300 |
52 | 0.43 | 66 | 100 |
56 | 0.36 | 59 | 100 |