The External Regenerant Installation Kit (P/N 038018) contains all of the components needed to install and operate the Dionex SC-CERS 500 with a pressurized water reservoir. The kit contains one 4-L bottle, one pressure regulator (0-30 psi/0-210 KPa) and appropriate tubing (P/N 039164).
Procedure
  1. Make the following air line connections:
  2. Locate the pieces of tinted 1/8" OD plastic tubing (P/N 050089) supplied in the Installation Parts Kit.
  3. Push the end of one piece of 1/8" OD tubing over the barbed fitting of the regulator. Connect the other end of the tubing to the source of air pressure.
  4. Push one end of the second piece of 1/8" OD tubing over the other barbed fitting of the regulator. Push the other end of this tubing over the barbed fitting (P/N 050077) in the pressure inlet of the plastic reservoir (see the figure below).
  5. Make the following water line connections.
  6. Use a coupler (P/N 039056) to connect one end of the 30" tubing assembly (P/N 035727) that comes in the Installation Kit to the water reservoir. Connect the other end of this tubing to the REGEN IN port of the Eluent Suppressor component of the Dionex SC-CERS 500.
  7. Using a coupler (P/N 039056) and a 1/8" OD piece of tubing (P/N 035728) from the Installation Kit, connect one end of this line to the waste port of the Dionex SC-CERS 500 and then connect the other end of the line to the Gas Separator Waste Tube.
  8. Fill the water source reservoir. Make sure that the O-ring is inside the cap of the reservoir before screwing the cap onto the reservoir. Screw the cap onto the reservoir tightly and place the reservoir near the Chromatography Module.
  9. With no current applied, use the air pressure gauge to adjust the external water flow rate to approximately 3-5 mL/ min for the Dionex SC-CERS 500 (4 mm) and 1-2 mL/min for the Dionex SC-CERS 500 (2 mm).
  10. The external water flow rate is controlled by two factors: the pressure applied to the water reservoir (0-25 psi) and the current setting on the Electrolytically Regenerated Suppressor Control unit. After the current is applied, the flow rate will drop due to gas formation in the regenerant chambers and the pressure must then be adjusted to give the correct flow rate at the specific current setting required for the application.
  11. See AutoSuppression External Water Mode.
AutoSuppression External Water Mode Plumbing Diagram
AutoSuppression External Water Mode Plumbing Diagram
NOTE
A safety relief valve on the reservoir regulator prevents pressure greater than 25 psi from being applied to the water reservoir.