The Dionex SC-CERS 500 comes pre-plumbed and the detailed plumbing schematic is shown in the following figure.
Procedure
- The column outlet should be connected to the line labeled “Column Out.”
- The line labeled “Cell in” should be connected to the conductivity cell inlet.
- Do not connect any backpressure coils or additional tubing that may add backpressure to the conductivity cell. Remove all preinstalled tubing and backpressure coils.
- The line from the conductivity cell should be connected using a coupler to an 1/8" waste line and diverted to waste.
CAUTION
No backpressure is required for the Dionex SC-CERS 500 suppressor. Remove any backpressure coils, if present.