Sum of Eluent Concentration Calculation
When using the Dionex SC-CERS 500 (4 mm) and Dionex SC-CERS 500 (2 mm), the sum of the eluent concentration can be calculated using the equation below:
Sum of eluent concentration (mN) = MSA (mM)