The following equations determine the correct amount of current that should be applied to a Dionex SC-CERS 500 based on the eluent concentration (mN) and flow rate (mL/min) for an application.
Dionex SC-CERS 500 (4 mm)
Operating Current (mA) = sum of eluent concentration (mN) * flow rate (mL/min) * 2.924
Dionex SC-CERS 500 (2 mm)
Operating Current (mA) = sum of eluent concentration (mN) * flow rate (mL/min) * 2.924