To ensure the optimal performance of the Dionex SC-CERS 500, avoid applying excess current to the suppressors. Excess operation currents lead to excess heat generation, which can reduce the suppressor lifetime and cause higher chromatographic baseline noise.
Dionex SC-CERS 500 (4 mm) – Do not exceed 300 mA
Maximum Current (mA) = sum of eluent concentration (mN) * flow rate (ml/min) * 4
Dionex SC-CERS 500 (2 mm) – Do not exceed 100 mA
Maximum Current (mA) = sum of eluent concentration (mN) * flow rate (ml/min) * 4