The Dionex SC-CERS 500 comes pre-plumbed, and the detailed plumbing schematic is shown in the figure below.
Procedure
- The column outlet should be connected to the line labeled “Column Out.”
- The line labeled “Cell in” should be connected to the conductivity cell inlet.
- Do not connect any backpressure coils or additional tubing that may add backpressure to the conductivity cell. Remove all preinstalled tubing and backpressure coils.
- The line from the conductivity cell should be connected using a coupler to an 1/8" line labeled “Cell Out”. An 1/8" waste line should be connected to the port labeled “To Waste.” Do not use 1/16" black/colored tubing for the waste line because this tubing adds backpressure to the device.
- Install a gas separator waste tube at the end of the waste line.