Depending on the specific components (analytical column, conductivity cell) in the system, 1/4-28 or 10-32 ferrule/bolt liquid lines may be required. All necessary tubing and fittings are supplied in the system, detector, Dionex RFC-10 or Dionex RFC-30 Ship Kits.
To purchase or assemble 1/4-28 or 10-32 ferrule/bolt liquid lines, refer to, “DIONEX Liquid Line Fittings” (P/N 031432). Always use 0.005" ID PEEK tubing with 10-32 ferrule/bolt fittings on 2 mm systems. Use 0.010" ID PEEK tubing with 10-32 ferrule/bolt fittings on 4 mm systems.
Install the Dionex SC-CERS 500 in the Chromatography Module as described in Installation.