The AutoSuppression Recycle Mode is the easiest method of operation. As the eluent passes through the suppressor, it is substantially neutralized to produce its weakly ionized form.
After passing through the conductivity cell, this effluent is redirected to the regenerant inlet on the ES component, thus supplying it with a source of water containing a small amount of dilute MSA and diluted analyte (see the following figure). The main advantage of this mode is its simplicity and ease of use. It is not necessary to have an external supply of water available for the suppressor. Refer to Select an operation mode to select the appropriate mode of operation for your application.
CAUTION
Only use the AutoSuppression Recycle Mode for MSA eluents without organic solvents.
AutoSuppression Recycle Mode Plumbing Diagram
AutoSuppression Recycle Mode Plumbing Diagram