Installing the Dionex SC-CERS 500 - Installing the Dionex SC-CERS 500 - SC-CERS 500 Suppressor User Guide - SC-CERS 500 - Installing the Dionex SC-CERS 500 - SC-CERS 500 - topic
SC-CERS 500 Suppressor User Guide
- Product Tree
- Chromatography > Ion Chromatography > Suppressors > SC-CERS 500
- Document Type
- User Guide
- Remove all existing backpressure coils and long lengths of tubing leading into the conductivity cell.
- No backpressure coils are required for the Dionex SC-CERS 500 suppressor operation.
- Assemble and install the Gas Separator Waste Tube and the waste line
- Install the two regenerated Dionex SC-CERS 500 components (ES and AC components) using the lines and fittings shipped with the Dionex SC-CERS 500.