The existing SRS/ERS power supply can be used for powering the ES component of the Dionex SC-CERS 500 device. Choose SRS or ERS as the suppressor option when operating from Chromeleon, PeakNet, or from the front panel of the instrument.
The ES Component is an electrolytic device and will operate using any SRS/ERS power supply. The AC Component is a chemical suppressor and hence requires no power.
Always use the recommended currents for a given application. Calculate the optimum current for your application as described in Dionex SC-CERS 500 current settings and set this current using the ED50A, CD25A or Dionex RFC-10/RFC-30.
When using older equipment, choose the lowest current available. For example, for 4 mm applications (such as 20 mM MSA with CS12A columns or 26 mM MSA with CS16 columns) use 100 mA setting and for 2 mm applications use 50 mA setting. For other eluent strengths of MSA refer to Dionex SC-CERS 500 current settings for calculating the optimized current setting.