Refer to AutoSuppression Recycle Mode, Using a Dionex RFC-10 or Dionex RFC-30 power supply to power the Dionex SC-CERS 500, and AutoSuppression External Water Mode for recommended current settings required for the chromatographic conditions required in the specific application being performed.
Procedure
- On CD20/25/25A, ED50/50A and ED40 detectors in Dionex DX-500, Dionex DX-600, and Dionex DX-320 instruments, use the Select keys to navigate to the SRS field in the Main Menu, then change the current setting.
- The read-out is the approximate current level.
Some general guidelines for setting the current are noted below.
- The lower the concentration of the eluent, the lower the current needed for total suppression. This depends on both the nature of the eluent components and their concentration.
- The lower the eluent flow rate, the lower the current required.
- Too high a current may create unnecessary baseline noise and reduce suppressor lifetime.
- Less current is required for Dionex SC-CERS 500 (2 mm) than Dionex SC-CERS 500 (4 mm).
- If background increases over time at a given current setting for a given application, increase current setting (see High background conductivity). The maximum recommended setting for the Dionex SC-CERS 500 (4 mm) is 300 mA. The maximum recommended setting for the Dionex SC-CERS 500 (2 mm) is 100 mA.
- If solvent is used in the eluent, use the current setting recommended for electrolytic suppression of aqueous eluents.