The Gas Separator Waste Tube (P/N 045460) is an integral part of the Dionex SC-CERS 500, similar to the Cation Electrolytically Regenerated Suppressor (CERS 500) system. Its function is to ensure the separation of any hydrogen/oxygen gas generated in the Dionex SC-CERS 500 during the electrolytic operation.
The Gas Separator Waste Tube is used to avoid concentrating the gas in the waste container. The gas separator waste tube is shipped in the ED40, ED50, ED50A, CD20, CD25, CD25A, Dionex DX-320, and Dionex RFC-30 Ship Kits.
CAUTION
Do not cap the waste reservoir. The very small amount of hydrogen gas generated by the Dionex SC-CERS 500 is not dangerous unless the gas is trapped in a closed container and allowed to concentrate. The gas separator waste tube must be open to the atmosphere and not in a confined space to operate properly.
Procedure
  1. Use one or two couplers (P/N 045463) to connect two or three lengths of 1/2" ID black polyethylene tubing (P/N 045462), depending of the depth of your waste container.
  2. It is important that the top of the waste separator tube extend above the top of the waste container as shown in the following figure.
  3. Place the Gas Separator Waste Tube with the 1/8" OD tubing attached into the waste container. Ensure the following:
  4. The bottom of the gas separator waste tube is resting on the floor of the waste container.
  5. The top of the device (where the white 1/8" OD tubing meets the black 1/2" OD tubing) is above the top of the container.
  6. The gas separator waste tube and the waste container are open to the atmosphere.
  7. AutoSuppression Recycle Mode Plumbing Diagram
    AutoSuppression Recycle Mode Plumbing Diagram