No backpressure coils are required for the Dionex SC-CERS 500 Suppressor operation. Remove any existing backpressure coils before installing the Dionex SC-CERS 500.
No backpressure coils are required for the Dionex SC-CERS 500 Suppressor operation. Remove any existing backpressure coils before installing the Dionex SC-CERS 500.