The Dionex SC-CERS 500 is designed to be run on any Dionex Ion Chromatograph (IC) equipped with a cation exchange column set and suppressed conductivity detection. See the following table for details.
Dionex Ion Chromatograph Series Module |
| Dionex RFC-10 or Dionex RFC-30 Suppressor Controller Required? |
|
|---|---|---|---|
2000i |
| YES |
|
QIC |
| YES |
|
2000 SP |
| YES |
|
4000i |
| YES |
|
4500i |
| YES |
|
8000 |
| YES |
|
8100 | CDM-2, PED CDM-3, PED-2 | YES RECOMMENDED |
(for 1 mA increment control) |
8200 | CDM-3, PED-2 | RECOMMENDED | (for 1 mA increment control) |
Dionex DX-320 | IC20 IC25 IC25A | RECOMMENDED RECOMMENDED NO | (for 1 mA increment control) (for 1 mA increment control) (integral with detector) |
Dionex DX-300 | CDM-2, PED CDM-3, PED-2 | YES RECOMMENDED |
(for 1 mA increment control) |
Dionex DX-500 |
| RECOMMENDED | (for 1 mA increment control) |
Dionex DX-600 | CD25 CD25A ED50 ED50A | RECOMMENDED NO RECOMMENDED NO | (for 1 mA increment control) (integral with detector) (for 1 mA increment control) (integral with detector) |
Dionex DX-800 |
| RECOMMENDED | (for 1 mA increment control) |
Dionex ICS-1000/1500/2000 /1100/1600/2100 |
| NO | (integral with system) |
Dionex ICS-2500 |
| NO | (integral with detector) |
Dionex ICS-3000 /5000/5000+ |
| NO | (integral with detector) |
The Dionex SC-CERS 500 is installed in the column compartment of the chromatography module immediately after the analytical column and before the conductivity detector cell. The mounting location depends on the system. See the following table.
System | Mounting location |
|---|---|
Dionex ICS-5000+/5000/3000 | SC-CERS 500 ES: On the CD in the DC module |
SC-CERS 500 AS: On the suppressor plate of the second detector console (if fitted), or in the top section of the upper compartment. | |
Dionex ICS-2100/2000/1600/1500/1100/1000 | SC-CERS 500 ES: On the vertical suppressor brackets between the CD cell and column in the component panel. |
SC-CERS 500 AS: On the horizontal suppressor brackets at the top of the component panel. | |
Dionex Integrion | SC-CERS 500 ES: On the tangs in the detector compartment. |
SC-CERS 500 AS: Affixed next to the ES in the detector compartment using double sided tape or hook and loop. |