The following components are required for the installation of the Dionex SC-CERS 500 in a Dionex Ion Chromatograph.
| P/N | Description |
|---|---|---|
Found in: | 046297 046298 045935 050130 059218 045720 | ED40, ED50/A Shipkit CD20, CD25/A Shipkit CDM-3 On-Line Shipkit Dionex DX-120 Shipkit Dionex DX-320 Shipkit RFC-30 Shipkit |
Additional components are required to plumb a 2-mm system: | 052324 | Microbore tubing (containing tubing, fittings and 2-mm backpressure coils required for plumbing a 2-mm system). NOTE: Backpressure coils must not be used with Dionex SC-CERS 500. |
SC-CERS 500 Spare Parts: | 016388 | Syringe, 1.0 mL, disposable For flushing the Dionex SC-CERS 500 at start-up. |
024305 | Syringe Adapter, female Luer lock, 1/4-28 threads | |
Installation Kit for LC30: | 060044 | Metal bracket, spacers and screws required for mounting the SC-CERS 500 AC in a LC30. |
Options: Pressurized Water Delivery System (used with the AutoSuppression External Water Mode) | 038018 | External Regenerant Installation Kit For Dionex SC-CERS 500 suppressor operation in the External Water mode when up to 40% HPLC solvents are used in the eluent. Contains one pressure regulator (0-30 psi/0-210 kPa), and all tubing and fittings required to install the Dionex SC-CERS 500. |
RFC-10 / RFC-30 |
| See the Reagent-Free Controller Manual for detailed instructions. |
CAUTION
The Dionex SC-CERS 500 must be operated with the gas separator was tube (P/N 045460).
NOTE
During the course of installing and using the Dionex SC-CERS 500, it may be necessary to assemble 1/4-28 or 10-32 ferrule/ bolt style liquid lines. See, “Installation of DIONEX Liquid Line Fittings,” Document No. 031432, for complete details.