The Dionex Salt Converter-Cation Electrolytically Regenerated Suppressor 500 (Dionex SC-CERS 500) contains components that are heat sensitive. The suppressor should not be subjected to temperatures above 40°C during shipment, storage, or operation.
Ensure the suppressor is stored in a temperature controlled environment away from direct exposure to sunlight or other sources of heat. Do not store the suppressor in a non-temperature controlled environment where temperatures exceed 40°C, such as a parked car, a tool shed, or a lab-bench in close proximity to an open window with direct sunlight.