The AutoSuppression External Water Mode is used for any applications requiring organic solvents in the eluent. This mode uses a constant source of deionized water from a pressurized bottle or a line source of deionized water that delivers at least 3 to 5 mL/ min. The amount of water flowing through the regenerant chambers is therefore independent of the eluent flow rate.
The AutoSuppression External Water Mode eliminates the potential for buildup of contaminating ions resulting from the oxidation of solvents. If it is determined that the system must be plumbed for applications requiring AutoSuppression External Water Mode, any analysis performed using the AutoSuppression Recycle Mode can also be performed using the AutoSuppression External Water Mode.
See Operation for more information.