The Dionex SC-CERS 500 is a post column electrolytic eluent suppressor for cation exchange applications using conductivity detection.
The Dionex SC-CERS 500 is used in cation exchange applications as a replacement for the Dionex CERS 500 when extended linearity or increased sensitivity for ammonium and amines are required. The Dionex SC-CERS 500 converts weakly ionized amines and ammonium to the highly ionized methanesulfonate acid, thus increasing their response and extending the linear range to three orders of magnitude.
The Dionex SC-CERS 500 is a specialty suppressor package that consists of:
- An Eluent Suppressor component (ES)
- An Analyte Converter component (AC)
This enables the suppressed conductivity detection of low-level ammonia and other amines with an extended linear response.
The Dionex SC-CERS 500 is installed in a cation ion exchange system and uses the same power supply used for SRS and ERS suppressors, as shown in the following figure.
In the Eluent Suppressor (ES) component, the majority of the eluent (methanesulfonic acid, H+MSA–) is suppressed, but the analytes remain in the MSA– salt form; for example, Na+MSA– in a background of dilute methanesulfonic acid.
In the Analyte Converter (AC) component, the analyte cation is exchanged for a hydronium ion (H+), thus resulting in conversion of the analyte to methanesulfonic acid (H+ MSA–). The analyte is detected as methanesulfonic acid in a background of methanesulfonic acid. See the following figure.
For example, ammonium hydroxide (ammonia) which is weakly dissociated is converted to ammonium MSA in the ES component, and in the AC component the ammonium MSA is converted to the conductive MSA form. Since MSA is fully dissociated, linear response vs. concentration calibration is observed at a wide range of concentration.
The Dionex SC-CERS 500 suppressor is recommended and optimized for isocratic MSA (methanesulfonic acid) only. The Dionex SC-CERS 500 is compatible with typical HPLC solvents up to 40%. The external water mode must be used for eluents containing solvents.
The Dionex SC-CERS 500 is available in both 4 mm and 2 mm formats. The Dionex SC-CERS 500 (2 mm) is specially designed with reduced internal volume to ensure optimum performance with 2 mm and 3 mm i. d. columns and microbore systems.
The electrolytic suppressor component of the Dionex SC-CERS 500 includes two regenerant compartments and one eluent compartment separated by ion exchange membranes. Regenerant flow channels and an eluent flow channel are defined by the membranes. The eluent flow is in a direction that is concurrent to the regenerant flow in the Eluent Suppressor component and countercurrent in the Analyte Converter.
In the Eluent Suppressor, electrodes are placed along the regenerant channels. When an electrical potential is applied across the electrodes, water from the regenerant channels is electrolyzed, supplying regenerant hydroxide ions (OH-) for the neutralization reaction. The membrane allows these hydroxide ions to pass into the eluent chamber resulting in the conversion of the electrolyte of the eluent to a weakly ionized form. Eluent anions are simultaneously passed into the regenerant chamber to maintain charge balance.
In the Eluent Suppressor, the electrode lengths are optimized to promote a transfer of methane sulfonic acid across the membranes at the outlet end. This transfer forms dilute MSA in the eluent stream resulting in a slightly elevated background and the analyte is in the MSA form.
The Analyte Converter is a chemical suppression system and has no electrodes. This component exchanges the analyte for hydronium ions, forming methanesulfonic acid.
NOTE
Assistance is available for any problem that may be encountered during the shipment or operation of Thermo Scientific Dionex instrumentation and columns through the Thermo Scientific North America Technical Call Center at 1-800-DIONEX-0 (1-800-346-6390) or through any of the Thermo Scientific Offices.