If you have an Ardia server in your laboratory, you may register an Ardia server to Proteome Discoverer. Once registered to an Ardia, you may load remote injections into studies and publish results from studies to the Ardia server. You can also register a single Proteome Discoverer installation for automated analysis. For more information, refer to the Proteome Discoverer User Guide.