New Features in Proteome Discoverer 3.2 - Proteome Discoverer Software - Proteome Discoverer Software 3.2 Release Notes - Features - Proteome Discoverer 3.2 Release Notes - New Features in Proteome Discoverer 3.2 - Proteome Discoverer Software - Proteome Discoverer Software
Proteome Discoverer Software 3.2 Release Notes
- Product Tree
- Desktop Software > Proteome Discoverer Software
- Ardia Platform > Software > Proteome Discoverer Software
- Document Type
- Release Notes
- Software version
- 3.2
- CHIMERYS™ 4.0: The CHIMERYS intelligent search includes several new improvements:
- - 2x reduction in analysis time
- - Support for new PTMs: acetylation, ubiquitinylation (K-GG), unmodified cysteine, and preOmics cysteine modification
- - Improved PTM localization for DIA data
- - Apex and area quantification
- - Support for larger datasets using Quan in All Files (approximately 300 GB for cloud-based CHIMERYS and 500 GB for CHIMERYS on Ardia Server)
- - Semi-tryptic digest support
- INFERYS™ 4.7: New PTMs (see new CHIMERYS features listed above)
- New tables for the display of DIA and other data
- - DIA Precursors
- - DIA Precursor Groups
- - Peptide Sequence Groups
- TMTpro™ 32plex and TMTpro 35plex
- - New Reporter Ion Control Channels Normalizer node
- - New default TMT 32plex and TMT 35plex methods
- - New default TMT 32plex and TMT 35plex workflows
- Annotation of proteins, peptide groups, PSMs, and MS/MS spectra in the Ardia™ Platform Data Explorer application and Object Picker for PD results uploaded to Ardia via the Publish to Ardia or automated analysis features
- New Statistical Insights node to provide custom summary statistics
- Additional protein group level FDR