Suggested recovery actions - Suggested recovery actions - Proteome Discoverer 3.3 Release Notes - Proteome Discoverer Software - Proteome Discoverer 3.3 Release Notes - Suggested recovery actions - Known Issues - Proteome Discoverer 3.3 Release Notes - Proteome Discoverer Software - Proteome Discoverer Software - topic
Proteome Discoverer 3.3 Release Notes
- Product Tree
- Ardia Platform > Software > Proteome Discoverer Software
- Desktop Software > Proteome Discoverer Software
- Document Type
- Release Notes
- Software version
- 3.3
- For some issues, restarting the application is the appropriate recovery action.
- In some cases (particularly issues that arise during data acquisition), restarting applications such as the Home Page might not ensure complete recovery. Typically, restarting the PC resolves issues, but some devices with error conditions might require power cycling.
- As a fix we generally do not recommend reinstalling the software or the operating system, which more commonly occurs after you install a new hard drive.