The following table lists defects that were resolved between Proteome Discoverer 3.2 and Proteome Discoverer 3.3. The table excludes Help issues and any cosmetic fixes. In some cases, the original abstract has been amended or expanded upon to better describe the reported issue. Both an engineering fix and follow-up testing (verified by our product evaluation department) have resolved each of these issues.
Item ID
Software section
Abstract
207794
Help
Documentation: A better description for INFERYS and spectral library prediction functionality is needed.
792818
Reporter ion quantification
TMT 32plex: Some values in the PSM are duplicated in the quan channel view plot.
842421
Reporter ion quantification
When using the precursor detector and reporter quantifier, the quan spectra are used for PSMs with high isolation interference.
93039
Normalization
Enable users to normalize only features found in all sample groups in quan analysis.
851668
Ardia
When users are not registered on any Ardia server: For studies containing many analysis files, the repeated check of remote files might slow down the performance.
842518
PSM filtering
Low-confident PSMs are still present when they belong to a high-confident peptide group.
824138
Spectral libraries
Predict from a FASTA file: The Activation Type parameter always shows HCD even when the CID option is selected during library creation.
838825
Spectrum selector
The workflow crashes when the Spectrum Selector node tries to store traces for the .raw files.
794155
Familiarization documentation
Familiarization Guide: In Chapter 2 for reporter ion quantification with phosphorylation, the Peptide Groups data source needs to be removed from the Volcano Plot steps.
798301
Fragment ions quantifier
The numbering index of the parameters is not correct in the Fragment Ions Quantifier node.
802042
Spectrum view
The Fragment Chart displays only charge 1 for DIA spectra.
802419
Protein Annotation node
When the database version changed, the workflow's annotation node and search engine node do not recognize the new version of the database, and the workflows need to be updated.
806818
MSPepSearch
MSPepSearch InforHint displays an incorrect warning about RAM memory limitations.
277263
Reporter ion quantification
An incorrect warning that reporter ion quantification cannot rescale abundance columns displays.
562597
Dependencies
The application does not use SQLite 118.
647791
Magellan server
Local databases (for example, Server DB) are insecure and not encrypted. Users can compromise settings and data integrity is not enforced.
746695
Percolator
Update to Percolator 3-07.
752141
Study
The word “Hints” does not appear in the the Execute State column in the analysis result table for jobs that completed with hints.
756957
CHIMERYS communication tester
Verify Discoverer can get an object from S3 and there are no network misconfigurations on the client side.
763521
FASTA file download
A FASTA file with and without subtaxonomies with the same TaxId cannot be downloaded.
772816
Protein table
The values of the #Protein Unique Peptides column in the Proteins table are 0.
777000
Ardia
If the default browser is not set to Microsoft Edge, then the sign in to Ardia server will not work.
778296
Processing
Processing workflow failed: Caller is unauthorized to delete specified resource[0].
782322
CHIMERYS
The CHIMERYS node failed because it cannot process spectra.
758266
Export
Astral .raw files exported to .mgf files contain no precursor mass in cases where the precursor detector is used.
856182
Reporter ion quantification
TMT quan values might be swapped when no sample grouping is used.
856230
Label free quan results
When the Precursor Ions Quantifier node is used, the chromatogram for the DIA Precursors table is missing.
764341
Plots
Originally,hidden tables (even after being made visible) cannot be used to plot histograms.
797992
CHIMERYS on Ardia
A gateway timeout occurred when submitting a job that resulted in a conflict.
829338
INFERYS library prediction, export, and MSPepSearch
Non-target peptides are added to the predicted library as targets when predicting from a protein FASTA file. The results failed MSPepSearch searches using a target/decoy due to insufficient target/decoys needed for FDR.
799390
CHIMERYS cloud
CHIMERYS skips the final merge step and downloads results for input files separately, reducing the length of time to finalize the result.
802042
Spectrum view
Only singly charged fragments are annotated for DIA spectra.