You can remove CHIMERYS inclusion files.
Procedure
  1. Do one of the following:
  2. Select Administration > Maintain CHIMERYS Inclusion Files.
  3. Select CHIMERYS Inclusion Files .
  4. The CHIMERYS Inclusion Files page opens.
  5. Select the inclusion file that you want to remove.
  6. Select Remove.