The following table describes the Percolator configuration parameters.
Parameter | Description |
|---|---|
Performance | |
Max. # Parallel Input File Creations | Maximum number of operations that create the input files for Percolator. The application needs to be restarted in order to let changes to this setting take effect. A number high than the number of logical processors available to the application will automatically be limited accordingly. Minimum value = 1, Maximum value = 12 |
Max. # Parallel Executions | Maximum number of Percolator processes that can be started in parallel when executing multiple workflows. The application needs to be restarted in order to let changes to this setting take effect. A number high than the number of logical processors available to the application will automatically be limited accordingly. Minimum value = 1, Maximum value = 12 |