The following table describes the configuration parameters for the INFERYS Spectrum Library Prediction.
Parameter | Description |
|---|---|
Service Configuration | |
Max. Entries in Sequence Database | The maximum number of sequences in the sequence database provided for library prediction. Minimum value = 1, Maximum value = 200000 |