The following table describes the configuration parameters for the Comet node.
Parameter | Definition |
|---|---|
Default Confidence Thresholds | |
High Confidence Threshold | Peptide matches with a score higher than the given value are assumed as high confident. Minimum value = 0, Maximum value = (unchecked) |
Medium Confidence Threshold | Peptide matches with a score higher than the given value are assumed as medium confident. Minimum value = 0, Maximum value = (unchecked) |