You can load remote files from the Ardia Advanced Tower Server directly into a study in the Proteome Discoverer application. When a file is imported from the Ardia Advanced Tower Server into a study, a green check mark appears in the Remote Status column. For any files imported locally, a green check mark appears in the Local Status column.
During the analysis, the Proteome Discoverer application downloads the remote injection from the Ardia Advanced Tower Server to the local processing computer and run it.
The analysis is completed locally and results are also stored locally until you publish them to the Ardia Advanced Tower Server. See Publish analyses to the Ardia Platform.
NOTE
You can also add existing MSF files to a study. The application adds the MSF file as a new result to the Analysis Results page and adds the raw data files that were processed to create the MSF files to the Input Files page.
For more information on the Ardia Repository parameters, see Ardia Repository parameters in the Ardia Platform Help portal.
Prerequisites
  1. You have opened a study in the Proteome Discoverer application.
Procedure
  1. To add remote results to a study, do the following:
  2. After the study opens, select the Input Files page in the study.
  3. Select the Add Files dropdown menu.
  4. Select Add Remote Results.
  5. The Pick Documents dialog box opens.
  6. In the Ardia Repository, locate your folder.
  7. Select the file(s) you want to add.
  8. Select Open.
  9. The file appears on the Input Files page. There is a green check mark in the Remote Status column.
  10. You can now create your analysis.
  11. To add remote fractions to a study, do the following:
  12. After the study opens, select the Input Files page in the study.
  13. Select the Add Files dropdown menu.
  14. Select Add Remote Fractions.
  15. The Pick Injections dialog box opens.
  16. In the Ardia Repository, locate your folder.
  17. Select the file(s) you want to add.
  18. Select Open.
  19. The file appears on the Input Files page. There is a green check mark in the Remote Status column.
  20. To add remote injections to a study, do the following:
  21. After the study opens, select the Input Files page in the study.
  22. Select the Add Files dropdown menu.
  23. Select Add Remote Injections.
  24. The Pick Injections dialog box opens.
  25. In the Ardia Repository, locate your folder.
  26. Select the file(s) you want to add.
  27. Select Open.
  28. The file appears on the Input Files page. There is a green check mark in the Remote Status column.