The following table describes the parameters for the PCM Grouper node.
Parameter | Definition |
|---|---|
Site Probability Threshold | Specifies that modifications with a lower site-probability than the specified threshold are not shown for the peptides. Minimum value = 25.0; Maximum value = 100.0 |
Split by File | If set to True, PCM Groups are split by file. |