The following table describes the parameters for the Fragment Ions Quantifier node.
Fragment Ions Quantifier node parameters
Parameter
Definition
Peptides to Use
Specifies the peptides to use for protein quantification.
  • Unique: Uses only peptides that are not shared among different proteins or protein groups for protein quantification.
  • (Default) Unique + Razor: Uses all quantification results that are not shared among different peptide groups for protein quantification. All shared quantification results are used for one peptide group only.
  • All: Uses all peptides for protein quantification, whether or not they are unique.
This parameter adds the # Razor Peptides column to the Proteins page of the result report when you use razor peptides for quantification.
Consider Protein Groups for Peptide Uniqueness
Defines peptide uniqueness on the basis of protein groups rather than individual proteins.
  • (Default) True: Considers a peptide unique if it is included in only one protein group.
  • False: Considers a peptide unique if it is included in only one protein.
For best results, use the default parameter. When you use larger protein (FASTA) databases, peptides commonly appear in multiple homologous proteins. If you set this parameter to False, the results might not contain many quantification results.
Normalization Mode
Specifies how to perform normalization to correct for experimental bias.
  • (Default) None: Does not apply any normalization.
  • Total Peptide Amount: After aggregating all abundance values per quantification channel, the node normalizes the abundance values of each channel with a constant channel-specific factor so that all channels have the same total peptide abundance at the end.
  • Specific Protein Amount: Calculates the normalization factor from the abundances of selected proteins in the specified FASTA file. You can specify a FASTA file (use the Proteins for Normalization parameter to specify the name of the FASTA file) that can contain one or multiple proteins. The Precursor Ions Quantifier node uses all proteins in the FASTA file that are contained in the result file and that have any protein abundance. It calculates the maximum sum for each file. The normalization factor is the factor of the sum of the sample and the maximum sum in the same file.
  • When normalization is based on specific proteins, the node sums the abundance values of all proteins in the result file that are contained in the selected FASTA file. It compares the protein sequences in the FASTA file to the sequences in the result file, not to the title lines. You can therefore use the same FASTA file when the title line is slightly different.
  • The node sums up the abundances for each sample. Then it calculates the normalization factor on the basis of these abundance sums.
  • If you set the Normalization Mode parameter to Specific Protein Amount but do not select a FASTA file, or if the result file contains no proteins that appear in the FASTA file, you cannot perform normalization.
  • RT Dependent: In this mode, the normalization is performed RT dependent by aligning sample abundances of consensus features from all peptides to the median abundance of all samples. This option is not supported for fractioned samples.
For more information on the Normalization Mode parameter, see Normalizing peptide groups and protein abundances.
Proteins for Normalization
Specifies the name of the FASTA database containing the proteins that the normalization should be based on. The database can contain a single protein sequence or multiple protein sequences.
This parameter is relevant only when you set the Normalization Mode parameter to Specific Protein Amount.
Scaling Mode
Determines how Precursor Ions Quantifier node scales abundances.
After it aggregates all the abundance values or optionally normalized abundance values per quantification channel, the node scales the abundance values of each channel as follows:
  • On All Average: For every protein and peptide in a file, the average of all samples is 100.
  • On Controls Average: For every protein and peptide in a file, the average of all control samples is 100. The node proportionally scales up or down all other channels by using the same factor. When you use multiplexed files, it processes samples from one file separately.
  • (Default) None: Does not scale abundances.
For Normalization
Specifies which peptides to use for deriving the normalization.
  • Use All Peptides: The normalization is derived by using all peptides.
  • Exclude Modified: The normalization is derived by only using peptides without the specified modifications.
  • Use Only Modified: The normalization is derived by exclusively using peptides with the specified modifications.
For Protein Roll-up
Specifies which peptides to use for calculation protein abundances.
  • Use All Peptides: The protein abundances are calculated by using all peptides.
  • Exclude Modified: The protein abundances are calculated by only using all peptides without the specified modifications.
  • Use Only Modified: The protein abundances are calculated by exclusively using peptides with the specified modifications.
For Pairwise Ratios
Specifies which peptides to use for calculation protein ratios based on the ‘Pairwise Ratio Based’ mode. The setting does not affect protein ratios directly calculated from protein abundances in the ‘Protein Abundance Based’ mode.
  • Use All Peptides: The protein ratios are calculated by using all peptides.
  • Exclude Modified: The protein ratios are calculated by only using peptides without the specified modifications.
  • Use Only Modified: The protein ratios are calculated by exclusively using peptides with the specified modifications.
1. Considered Peptide Modification
Peptides with the specified modification will be considered for inclusion or exclusion in protein ratios based on the other settings in this section.
2. Considered Peptide Modification
Peptides with the specified modification will be considered for inclusion or exclusion in protein ratios based on the other settings in this section.
3. Considered Peptide Modification
Peptides with the specified modification will be considered for inclusion or exclusion in protein ratios based on the other settings in this section.
N-Terminal Considered Peptide Modification
Peptides with the specified modification will be considered for inclusion or exclusion in protein ratios based on the other settings in this section.
C-Terminal Considered Peptide Modification
Peptides with the specified modification will be considered for inclusion or exclusion in protein ratios based on the other settings in this section.
N-Terminal Considered Protein Modification
Peptides with the specified modification will be considered for inclusion or exclusion in protein ratios based on the other settings in this section.
C-Terminal Considered Protein Modification
Peptides with the specified modification will be considered for inclusion or exclusion in protein ratios based on the other settings in this section.
Protein Abundance Calculation
Specifies how protein abundances are calculated.
  • Summed Abundances: Protein abundances are calculated by summing samples abundances of the connected peptide groups.
  • Top N Average: Protein abundances are calculated as the average of the N most abundant distinct peptide groups.
N for Top N
Specifies the number of distinct peptides used for calculating the protein abundance values from the connected peptides according to the Top N approach. The value is only relevant if ‘Top N Average’ is selected for calculating protein abundances.
Minimum = 1; Maximum = (unchecked)
Protein Ratio Calculation
Specifies how protein ratios are calculated.
  • Protein Abundance Based: Protein ratios are directly calculated from the grouped protein abundances.
  • Pairwise Ratio Based: Protein rations are calculated as the median of all possible pairwise peptide ratios calculated between replicates of all connected peptides.
Maximum Allowed Fold Change
Specifies the maximum fold change that can be reasonably expected for the observed ratios.
Reports the quantification ratios based on the maximum values. Values greater than the value selected by this option are replaced by the maximum or minimum value.
Range: 1–100 000; default: 100
With the default setting, ratios above 100 are set to 100, and ratios below 0.01 are set to 0.01.
Imputation Mode
Specifies how to treat missing values.
  • None: Does not impute any missing values.
  • Replicated Based Resampling: Imputes low-abundance values by using the already available method random sampling from the distribution of the lower fifth percentile of abundance values.
  • Low Abundance Resampling: Replaces missing values with random values sampled between the minimum and the lower 5% of all detected values.
  • (Default) k-Nearest Neighbors: Replaces missing values with the k-nearest neighbors detected among all samples.
For more information, see Impute missing values.
Hypothesis Test
Specifies how p-values are calculated for the reported quan ratios.
  • t-test (Background Based): t-test based on background population of proteins or peptides.
  • ANOVA (Individual Proteins): ANOVA based on the abundances of individual proteins or peptides. For this you need to specify that ratios are calculated based on protein abundances. In addition, you need at least three (biological) replicates in each condition of the ratio.
  • None: No hypothesis test is performed.
Calculate Hypothesis Test for Peptides
Specifies whether p-values are calculated for peptides.
1st Fold Change Threshold
Specifies the threshold for ratio changes to be highlighted as upregulated or downregulated. Ratios higher than the specified value or lower than 1/value are highlighted in the first highlight color. A ratio of 1.0 indicates no change.
Range: 1.0–1000.0; default: 2
2nd Fold Change Threshold
Specifies the threshold for ratio changes to be highlighted as upregulated or downregulated. Ratios higher than the specified value or lower than 1/value are highlighted in the second highlight color. A ratio of 1.0 means no change.
Range: 0.0–1000.0; default: 4
3rd Fold Change Threshold
Specifies the threshold for ratio changes to be highlighted as upregulated or downregulated. Ratios higher than the specified value or lower than 1/value are highlighted in the third highlight color. A ratio of 1.0 means no change.
Range: 0.0–1000.0; default: 6
4th Fold Change Threshold
Specifies the threshold for ratio changes to be highlighted as upregulated or downregulated. Ratios higher than the specified value or lower than 1/value are highlighted in the fourth highlight color.
Range: 0.0–1000.0; default: 8
5th Fold Change Threshold
Specifies the threshold for ratio changes to be highlighted as upregulated or downregulated. Ratios higher than the specified value or lower than 1/value are highlighted in the fifth highlight color.
Range: 0.0–1000.0; default: 10