The following table describes the PSM and Protein Grouper post-processing node
Parameter | Description |
|---|---|
Group PSMs as PCM | Group PSMs in peptide charge mass groups (PCMs). All items in the group are identical in sequence, theoretical mass, and charge. |
The following table describes the PSM and Protein Grouper post-processing node
Parameter | Description |
|---|---|
Group PSMs as PCM | Group PSMs in peptide charge mass groups (PCMs). All items in the group are identical in sequence, theoretical mass, and charge. |