The Proteome Discoverer application requires a license. In addition, ensure that the system meets the minimum requirements listed in the following table.
System | Minimum requirements | Recommended |
|---|---|---|
Computer |
|
|
Software |
| Same as minimum requirements |
MascotSM Server |
| Same as minimum requirements |
NOTE
Ensure that port 28199 is not blocked by firewalls.
NOTE
Before installing the Proteome Discoverer application, ensure that the Windows operating system has the Microsoft .NET Framework and Windows updates installed.