The Spectrum Selector node selects the subset of the set of spectra that you want to search. You must use this node with the Spectrum Files node.
When searches are based on MGF, MZDATA, or other external file formats, the extracted spectra might not have information about the activation type, mass analyzer, polarity, and so forth. However, certain processing nodes require some of this information. To enable such searches, you can use the Spectrum Selector node to provide these values.
NOTE
If you specify both a retention time range and a scan range, the application combines them into a single filter criterion; that is, it processes a scan only if it simultaneously falls within the specified retention time range and the specified scan range.
The following table describes the parameters for the Spectrum Selector node.
Parameters | Description |
|---|---|
Precursor Selection | Determines which precursor mass to use for a given MSn scan. This option applies only to higher-order MSn scans like MS3, MS4, and so forth.
|
Use Isotope Pattern in Precursor Reevaluation | Determines whether the application considers the isotope pattern in reevaluating precursors.
|
Provide Profile Spectra | Determines whether profile points should be provided. In automatic mode, the node checks whether there is any node in the workflow that requires profile spectra. |
Lower RT Limit | Specifies the lower retention-time limit. Retention time is the time in the mass chromatogram when any particular precursor ion is observed. The Lower RT Limit and the Upper RT Limit parameters set limits on the portion of the data that you want to analyze. Because very early and very late portions in the chromatogram typically do not contain the peptide ions of interest, you might want to analyze only that middle portion that contains useful data to save search time. If the value is 0.0, the application uses the lowest available retention time. Range: 0–no maximum; default: 0 |
Upper RT Limit | Specifies the upper retention-time limit. If the value is 0.0, the application uses the highest available retention time. Range: 0–no maximum; default: 0 |
First Scan | Specifies the scan number of the first scan to process. If you set this number to 0, the application uses the first available scan. Range: 0–no maximum; default: 0 |
Last Scan | Specifies the scan number of the last scan to process. If you set this number to 0, the application uses the last available scan. Range: 0–no maximum; default: 0 |
Ignore Specified Scans | Opens the Edit Parameter Text for Ignore Specified Scans dialog box so that you can enter a single scan or a range of scans to exclude from processing. To specify a single scan, enter its number in the dialog box. To specify a range, place a hyphen between the starting scan and the ending scan, for example, 2000–3000. For multiple entries, place a semicolon (;) between each entry, or place each entry on a separate line. |
Lowest Charge State | Filters out spectra with a precursor charge lower than the specified charge. Range: 0–no maximum; default: 0 |
Highest Charge State | Filters out spectra with a precursor charge higher than the specified charge. If you set the value to zero, there is no maximum value for the precursor charge. Range: 0–no maximum; default: 0 |
Min. Precursor Mass | Specifies the lower mass limit of a singly charged precursor ion to be processed. Range: 0.0–10 000.0 Da; default: 350 Da |
Max. Precursor Mass | Specifies the upper mass limit of a singly charged precursor ion to be processed. Sequest HT searches rely on indexed FASTA databases. The search engine considers only peptides that fall between 300 and 10000 as PSMs, so setting a value higher than 10000 does not result in the identification of more PSMs. If you set the value to zero, the value is set to the maximum precursor mass allowed. Range: 0.0–no maximum; default: 5000 Da |
Total Intensity Threshold | Specifies a minimum total intensity value and filters out tandem mass spectra that have a total intensity current (the sum of the intensities of all peaks in a spectrum) below the specified value. You can use this parameter to remove low-intensity data. Range 0.0–no maximum; default: 0 |
Minimum Peak Count | Specifies the minimum number of peaks in a tandem mass spectrum that is allowed to pass the filter and to be subjected to further processing in the workflow. Use this parameter to remove spectra that have a small number of peaks, because these typically produce low-quality hits. Range: 1–no maximum; default: 1 |
Mass Analyzer | Specifies the type of mass analyzer used to acquire the spectrum. This parameter filters out all but the type you are interested in. It is possible to have data from more than one type of mass analyzer in the same data input, for example, data from a hybrid instrument such as LTQ Orbitrap or LTQ FT. These are the available analyzers:
|
MS Order | Specifies the level of the mass spectrum to be processed, for example, MS/MS or MS3. Range: MS1–MS10; default: Is Not MS1 |
Activation Type | Specifies the fragmentation method used to generate the MS/MS spectra. The following activation types are available:
For descriptions, see Multidimensional separation experiments. |
Min. Collision Energy | Filters out the spectra of ions fragmented with low collision energy from the spectra of ions fragmented with high collision energy. This parameter filters out values below the specified value. Range: 0–100; default: 0 |
Max. Collision Energy | Filters out the spectra of ions fragmented with high collision energy from the spectra of ions fragmented with low collision energy. This option filters out values above the specified value. Range: 0–1000; default: 1000 |
Scan Type | Specifies the scan type of the spectrum. These are the available scan types:
|
Polarity Mode | Specifies the polarity of the spectrum:
|
MS1 Mass Range | This filter applies only to MS1 spectra. Those matching the specified mass range pass the filter. |
FAIMS CV | When the FAIMS CV value is unspecified (empty), the analysis ignores CV values and all spectra pass the filter. When you specify a CV value, the analysis processes only the input files that include spectra for this CV value. |
IMPORTANT The compound detection node requires MS1 spectra from every input file in an analysis, but it can process spectra for only one CV value per input file. To prevent an analysis with compound detection from failing, do the following:
| |
S/N Threshold (FT-only) | Specifies a signal-to-noise value below which peaks are removed. The S/N threshold has an effect on FT data only. Data acquired with other mass analyzers are not affected. Range 0.0–no maximum; default: 1.5 |
Unrecognized Charge Replacements | Specifies the charge states of the ions to be included in the search, if they are not in the scan header. If the charge state of the precursor cannot be determined, the spectrum is replicated for each of the specified precursor charges. Select from these settings:
If you select Automatic, the application applies a simple check for a +1 spectrum; otherwise, it generates a +2 and a +3 spectrum. |
Unrecognized Mass Analyzer Replacements | Specifies the mass spectrometer to use to produce the spectra, if it is not included in the scan header. Select from these settings:
|
Unrecognized MS Order Replacements | Specifies the MS order to use in case the application cannot retrieve it from the input. Range: MS1–MS10; default: MS2 |
Unrecognized Activation Type Replacements | Specifies the activation type to use in case the application cannot retrieve it from the input. Select one of these activation types:
|
Unrecognized Polarity Replacements | Specifies the polarity to use in case the application cannot retrieve it from the input.
|
Unrecognized MS Resolution@200 Replacements | Specifies the resolution to use at mass 200 for MS scans in case you cannot retrieve this information from the input. Range: 0–no maximum; default: 60 000 |
Unrecognized MSn Resolution@200 Replacements | Specifies the resolution to use at mass 200 for MS/MS scans in case you cannot retrieve this information from the input. Range: 0–no maximum; default: 30 000 |
Precursor Clipping Range Before | Specifies the lower limit below the isolation window of the mass range from which to extract peaks for the Precursor Isotope Pattern view, in Da. This view shows the m/z range of the corresponding master scan from which the fragmented precursor ion was extracted. For more information, see Use the Precursor Isotope Pattern view. The Precursor Clipping Range After specifies the upper limit of the m/z range. Range: 2.5–100.0 Da; default: 2.5 Da |
Precursor Clipping Range After | Specifies the upper limit above the isolation window of the mass range from which to extract peaks for the Precursor Isotope Pattern view, in Da. This view shows the m/z range of the corresponding master scan from which the fragmented precursor ion was extracted. For more information, see Use the Precursor Isotope Pattern view. The Precursor Clipping Range Before specifies the lower limit of the m/z range. Range: 5.5–100.0 Da; default: 5.5 Da |