Activate automatic FASTA index removal - Activate automatic FASTA index removal - Proteome Discoverer 3.1 User Guide - Proteome Discoverer Software - Activate automatic FASTA index removal - Proteome Discoverer Software - Proteome Discoverer Software - topic
Proteome Discoverer 3.1 User Guide
- Product Tree
- Ardia Platform > Software > Proteome Discoverer Software
- Desktop Software > Proteome Discoverer Software
- Document Type
- User Guide
- Software version
- 3.1
- Select the Auto Remove checkbox.
- Select Apply.
- In the Remove FASTA indexes confirmation box, select OK.