Activating your Ardia Proteome Discoverer license uses the same process as any other Proteome Discoverer license. See Managing the application licenses for instructions.
After you activate your Ardia Proteome Discoverer license, new menus and icons appear on the application tool bar, the study, and on the Administration page, such as the Ardia Server Management menu.
1234
No.DescriptionNo.Description
1
Ardia Server menu
3
Ardia Server Management menu
2
Open Remote Result icon
4
CHIMERYS on Ardia Server configuration