You can publish your analyses to the Ardia Platform. This process removes the analysis result file from the local processing computer and stores it on the Ardia Platform.
During the upload to the Ardia server, Proteome Discoverer creates a concatenated .pdResultx file that is stored on the server. This file contains multiple files, such as the PDRESULT file, the PDRESULTDETAILS file, the PDRESULTSDETAILSVIEW file, and the PDRESULTVIEW file.
If the analysis only contains a processing workflow (no consensus workflow), then Proteome Discoverer creates a concatenated .msfx file. This file contains multiple files, such as the MSF File, the MSFDETAILS File, the MSFDETAILSVIEW file, and MSFVIEW File.
Prerequisites
  1. You have completed a run and the analysis is saved locally.
Procedure
  1. Open a study.
  2. Select the Analysis Results page in the study.
  3. The Analysis Results page opens with a list of results.
  4. Select the result file that you want to publish to the Ardia Platform.
  5. Select Publish.
  6. The Select a Folder dialog box opens.
  7. Locate the folder that you want to publish to and select the checkbox of that folder.
  8. Select Open.
  9. An Upload Manager page opens in the Proteome Discoverer application with the status of the upload.
  10. You have the option to cancel the upload if necessary.
  11. After the upload is complete, in the status box for that file displays the following message: Upload finished.