After you register your Ardia CHIMERYS license, you must test the communication with the server.
Prerequisites
- You have registered your Ardia CHIMERYS license. See Configure the CHIMERYS search engine.
Procedure
- From the application menu bar, select Tools > CHIMERYS Communication Tester.
- The CHIMERYS Communication Tester and Job Monitoring Tool opens.
- Select the CHIMERYS Node dropdown menu and select CHIMERYS on Ardia Server.
- Select Test.
- If the test is successful, CHIMERYS on Ardia is ready to use.