To use the Ardia Server functionality, you must register your processing PC to the Ardia server.
Prerequisites
  1. You have activated your Proteome Discoverer license. See Managing the application licenses.
  2. You have activated your Ardia license for Proteome Discoverer. See Licensing the Ardia features in the Proteome Discoverer software.
Procedure
  1. On the Windows desktop, double-click Proteome Discoverer .
  2. The Start Page opens.
  3. Do one of the following:
  4. Select Ardia Server > Register on Ardia Server
  5. In the Ardia Server Management menu, select Server Registration.
  6. The Ardia Server Registration window opens.
  7. Select Register.
  8. The Ardia Platform Registration dialog box opens.
  9. In the Ardia Server URL box, enter the Ardia URL.
  10. Select Register.
  11. Close Proteome Discoverer and reopen the application.
  12. Proteome Discoverer automatically opens the Ardia Platform in a web browser with a single sign-on. See Sign in to the Ardia Platform.
  13. You can now open a study in the Proteome Discoverer application.
  14. (Optional) To adjust the number of download/upload attempts, do the following:
  15. Select Ardia Server > Server Registration.
  16. In the Configuration area, enter the number of download/upload attempts.
  17. NOTE
    The default is 3.