The following table describes the parameters in the FASTA Index Creator dialog box.
Parameters | Description |
|---|---|
General | Sets general options for the creation of the FASTA index. |
Auto Remove | Specifies whether the available FASTA indexes are removed from memory after the number of indexes reaches the specified maximum. The options are as follows:
|
Create Additional Decoy Database Index | Determines whether the application creates an index for the specified FASTA file and an index for the decoy version of the FASTA file. The options are as follows:
|
Input Data | Specifies the basic information that the application requires to create the index. |
FASTA File | Specifies the name of the FASTA database. |
Enzyme Name | Specifies the name of the enzyme used for digestion and the type of digestion. The type of digestion is listed in parentheses after the enzyme name, and can be one of these options:
|
Maximum Missed Cleavage Sites | Specifies the maximum number of internal cleavage sites (per peptide fragment) that an enzyme is allowed to miss when cleaving peptides during digestion. Range: 0–12; default: 2 |
Mass Range Settings | Sets the limits of the mass range of the singly charged precursor ion to be processed. |
Minimum Precursor Mass | Specifies the minimum mass of the precursor ion. Minimum: 0.0 Da; maximum: 10 000.0 Da; default: 350 Da |
Maximum Precursor Mass | Specifies the maximum mass of the precursor ion. Range: 0.0–10 000.0 Da; default: 5000 Da |
Use Average Precursor Mass | Determines whether the average mass is used to match the precursor ion. The options are as follows:
|
Static Modifications | Specifies the static modifications that occur on the amino acid. |
Peptide N-Terminus | Specifies the static modification that occurs on the N-terminus of the peptide. |
Peptide C-Terminus | Specifies the static modification that occurs on the C-terminus of the peptide. |
Static Modification boxes | Specifies the static modification that occurs anywhere on the amino acid side chain. |