The following table describes the License Manager command bar.
Command or feature | Description |
|---|---|
Activate | Opens the License Activation dialog box where you can enter a new activation code and activate the license on the current computer. |
Deactivate | Opens the License Deactivation dialog box where you can deactivate the software license. |
Scan for Missing Features | Activates a scan for newly installed processing workflow nodes. |
Show Expired Licenses | A checkbox that, when selected, displays any expired licenses. |