Hide ToolTips - Proteome Discoverer Software - Hide ToolTips - Proteome Discoverer Software - Proteome Discoverer Software
Proteome Discoverer 3.1 User Guide
- Product Tree
- Ardia Platform > Software > Proteome Discoverer Software
- Desktop Software > Proteome Discoverer Software
- Document Type
- User Guide
- Software version
- 3.1
- Right-click the empty space in the toolbar and select Customize.
- The Customize dialog box opens.
- Select the Options tab.
- To hide the ToolTips when you point to the icons, clear the Show ScreenTips on toolbars option.
- To save your changes and close the Customize dialog box, select Close.